Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIF4GD Monoclonal Antibody (OTI2H9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$231.00 - $516.29
Specifications
| Antigen | MIF4GD |
|---|---|
| Clone | OTI2H9 |
| Concentration | 0.68 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-254-0253
![]()
|
OriGene
TA504558S |
30 μL |
Each for $231.00
|
|
|||||
50-167-5216
![]()
|
OriGene
TA504558 |
100 μL |
Each for $516.29
|
|
|||||
Description
MIF4GD Monoclonal antibody specifically detects MIF4GD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MIF4GD | |
| 0.68 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| Primary |
| OTI2H9 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| IgG2b | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title