Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Migfilin Antibody (OTI3G1), HRP, Novus Biologicals™
SDP

Catalog No. NBP272691H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272691H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272691H Supplier Novus Biologicals Supplier No. NBP272691H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Migfilin Monoclonal specifically detects Migfilin in Human samples. It is validated for Western Blot.

Specifications

Antigen Migfilin
Applications Western Blot
Classification Monoclonal
Clone OTI3G1
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein
Gene Symbols FBLIM1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-242 of human Migfilin (NP_060026) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 54751.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.