Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Miglustat hydrochloride
GREENER_CHOICESDP

Catalog No. 311710 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-171-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 31-171-0 Supplier Tocris Bioscience™ Supplier No. 3117/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α-glucosidase I and II inhibitor; also inhibits ceramide-specific glycosyltransferases and exhibits broad spectrum antiviral activity

Miglustat hydrochloride is an orally active α-glucosidase I and II and ceramide-specific glycosyltransferase inhibitor. Rescues trafficking-deficient F508del-CFTR in human airway epithelial cells via inhibition of ER α-glucosidases I and II. Also has broad spectrum antiviral activity.

Chemical Identifiers

CAS 210110-90-0
Molecular Formula C10H22ClNO4

Specifications

Quantity 10 mg
Formula Weight 255.74
Purity >98%
Molecular Weight (g/mol) 255.739
Chemical Name or Material Miglustat hydrochloride
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.