Learn More
Eisco™ Milky Quartz Mineral Specimen
SCI_ED
Catalog No. S24710 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24710 1 Ea.
S26608 10/Pk.
S24711 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24710 Supplier Eisco™ Supplier No. ESNG0025
Add to Cart
Add to Cart
Milky Quartz Mineral Specimen has a tightly packed framework and hardness of 7, complete lack of cleavage planes, and distinct euhedral crystal habit.
  • For use in classroom identification labs and exercises
  • For use in lessons on mineral hardness, luster, and fracture
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Used in identification exercises, asking students to distinguish between it and other visually similar minerals such as feldspar or calcite
Specifications
Product Type Mineral
For Use With (Application) Lessons on mineral hardness, luster, and fracture
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Milky Quartz