Learn More
Honeywell™ Miller™ DuraFlex™ Python™ Harnesses
Catalog No. 19274636
Click to view available options
Buckle Style:
Mating
Mating/Tongue
Quick-Connect
D-Ring Location:
Back
Back, Side
Catalog No. 19-274-636
Supplier: Honeywell™ P9507/UGN
Combine stretchable DuraFlex torso straps with lightweight, cushioned shoulder straps for ultimate comfort
Come with an invisible protective coating that resists staining, repels dirt and grime, preserves color, and wipes clean in an instant, ensuring easy maintenance and longer service life.
  • Abrasion-resistant, green nylon webbing has contoured tubular edges and high-density foam cushioning
  • Does not bunch or kink, improving neck and shoulder comfort
  • Flexibility and ease of movement are enhanced by stretchable webbing at the torso, which reduces fatigue
  • Adjustable leg straps feature nonstretch webbing with a choice of tongue or mating buckles
  • Fully adjustable, tangle-free, and easy to don
  • All models feature a sliding back D-ring and pad
  • Universal size
  • Green
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; CSA Z259.10 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Material Nylon Webbing
Color Green
Buckle Style Mating
Buckle Style (Chest) Mating Chest Straps
D-Ring Location Back, Side
Buckle Style (Leg) Mating Leg Straps
Certifications/Compliance Meets all applicable OSHA, ANSI, and CSA requirements.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.