Learn More
Description
One-Piece Adjustable Mast
- Mast offset adjusts from 12 to 29 in. (30 to 74cm)
Three-Piece Portable Base
- Lightweight aluminum components; adjustable aluminum screws and a built-in level indicator
- No tools required for assembly
Floor Mount Sleeve
- Mounts to horizontal concrete or steel structures
Wall Mount Sleeve
- Mounts to vertical concrete or steel structures
Flush Floor Mount Sleeve
- Eliminates potential trip hazard
- Installs into 4 in. dia. (10cm) core hole
Vehicle Hitch Mount Sleeve
- Installs into 2 in. dia. (5cm) receiver on an attendant vehicle
- Provides portable anchor point
Specifications
Specifications
| Material | Aluminum |
| Load Bearing Capacity (Metric) | 204 kg |
| Load Bearing Capacity (English) | 450 lb. |
| Certifications/Compliance | OSHA, ANSI, CSA |
| Product Type | One piece adjustable mast |
| For Use With (Equipment) | DuraHoist confined space system |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.