Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Revolution™ DualTech™ Harnesses

Harnesses with DualTech webbing offering safety and functionality with features developed to meet key user needs

Specifications

Buckle Style (Leg) Tongue-Buckle Leg Straps
Certifications/Compliance OSHA, ANSI and CSA
Color Black
For Use With (Application) Construction, manufacturing and transportation fall protection
Style Full-body harness
View More Specs

Products 7
Catalog Number Mfr. No. D-Ring Location Includes Size Price Quantity & Availability  
Catalog Number Mfr. No. D-Ring Location Includes Size Price Quantity & Availability  
19-165-283
View Documents
Honeywell™
RDTFDTB/UBK
Front Integrated accessory system Universal N/A N/A N/A
19-165-285
View Documents
Honeywell™
RDTFDTB/3XLBK
Front Integrated accessory system 2X-Large/3X-Large N/A N/A N/A
19-165-289
View Documents
Honeywell™
RDTFDTBDP/UBK
Front, Side Pad, integrated accessory system Universal N/A N/A N/A
19-165-290
View Documents
Honeywell™
RDTFDTBDP/S/MBK
Front, Side Pad, integrated accessory system Small/Medium N/A N/A N/A
19-165-292
View Documents
Honeywell™
RDTFDTBBDP/UBK
Front, Side Removable belt and pad, integrated accessory system Universal N/A N/A N/A
19-165-294
View Documents
Honeywell™
RDTFDTBBDP/3XLBK
Front, Side Removable belt and pad, integrated accessory system 2X-Large/3X-Large N/A N/A N/A
19-165-302
View Documents
Honeywell™
RDTFDTBBDP/S/MBK
Front, Side Removable belt and pad, integrated accessory system Small/Medium N/A N/A N/A
Description

Description

  • Available with tongue buckle legs and front and/or side D-rings
  • Available with or without removable belt
  • Exceed worker expectations in comfort, fit, ease-of-use, style, durability, compliance, flexibility and convenience
Specifications

Specifications

Tongue-Buckle Leg Straps
Black
Full-body harness
OSHA, ANSI and CSA
Construction, manufacturing and transportation fall protection
DualTech™, Polyester
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.