Learn More
Description
- PivotLink™ connection with unique rotary design promotes greater comfort in bending/mobility; also serves as an attachment point for belts, tools and accessories as part of an integrated accessory system
- Cam buckle provides easy, one-handed adjustment/release and allows simultaneous adjustment of shoulder straps
- Semi-flexible ErgoArmor™ back shield minimizes impact from sharp/heavy snap hooks and self-retracting lifelines; knitted mesh membrane minimizes moisture absorption and maintains optimal air circulation
- Stand-up back D-ring simplifies connection
- DualTech™ webbing makes donning easy with two-sided, contrasting-color webbing and textures
- High-impact-resistant polymer web finials provide positive gripping surface for easy web adjustment
- Integrated self-contained label pack encapsulates labels, decreasing damage and loss
Order Info
Webbing tested to 7000 lb. tensile strength. Universal size.
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; CSA Z259.10 requirements.
Specifications
Specifications
| Color | Black |
| Material | DualTech Webbing |
| Product Type | Revolution Harness |
| Style | With Removable Belt |
| Size | Universal |
| For Use With (Application) | Construction, Fall Protection |
| Buckle Style (Leg) | Tongue-Buckle Leg Straps |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.