Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Miller™ StretchStop™ Lanyard with SofStop Shock Absorber
Combination of integral SofStop Shock Absorber with stretchable lanyard design
Catalog Number | Mfr. No. | Connector Types | Leg Configuration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Connector Types | Leg Configuration | Price | Quantity & Availability | ||||
19-034-896
|
Honeywell™
913RSSZ7/6FTGNC |
Locking Snap Hook and Locking Rebar Hook | Single-Leg | N/A | N/A | N/A | |||
19-066-225
|
Honeywell™
913SSZ7/6FTGN |
2 Locking snap hooks | Single-Leg | N/A | N/A | N/A | |||
Description
Choose from three options:
- 6-ft. single leg, 2 locking snap hooks
- ANSI Z359 Compliant, Two legs, locking snap hook and 2 locking rebar hooks (2.5 in.)
- ANSI Z359 Compliant, 6-ft. single leg, 1 locking snap hook and 1 locking rebar hook (2.5 in.)
Compliance
ANSI Z359 Compliant.
Specifications
Green | |
6 ft. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title