Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MINA Antibody (753002), Biotin, Novus Biologicals™
SDP

Catalog No. NB374575 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374575 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374575 Supplier Novus Biologicals Supplier No. FAB7476B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MINA Monoclonal antibody specifically detects MINA in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen MINA
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 753002
Conjugate Biotin
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias DKFZp762O1912, FLJ14393, MDIGDKFZp762O1912, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52
Host Species Mouse
Immunogen E. coli-derived recombinant human MINA, Met1-Gly192, Accession # Q8IUF8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 84864
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.