Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mind Bomb 1/MIB1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB164727 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164727 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164727 Supplier Novus Biologicals Supplier No. NBP281754AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mind Bomb 1/MIB1 Polyclonal antibody specifically detects Mind Bomb 1/MIB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Mind Bomb 1/MIB1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DAPK-interacting protein 1, DIP1, DIP-1FLJ90676, DKFZp686I0769, DKFZp761M1710, E3 ubiquitin-protein ligase MIB1, EC 6.3.2, EC 6.3.2.-, KIAA1323, MGC129660, MIB, Mind bomb homolog 1, mindbomb homolog 1 (Drosophila), ubiquitin ligase mind bomb, ubiquitin ligase protein MIB1, Zinc finger ZZ type with ankyrin repeat domain protein 2, ZZANK2MGC129659, ZZZ6
Host Species Rabbit
Immunogen MIB1 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human MIB1. The immunogen is located within amino acids 920 - 970 of MIB1. Amino Acid Squence: IPVLQKDKDNTNVNADVQK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 57534
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.