Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mind Bomb 2/MIB2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068224 Supplier Novus Biologicals Supplier No. NB10068224

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody has been used in 2 publications

Mind Bomb 2/MIB2 Polyclonal specifically detects Mind Bomb 2/MIB2 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).

Specifications

Antigen Mind Bomb 2/MIB2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Immunoprecipitation (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_543151.1
Gene Alias E3 ubiquitin-protein ligase MIB2, EC 6.3.2, EC 6.3.2.-, EC 6.3.2.13, FLJ20648, FLJ41406, Mind bomb homolog 2, mindbomb homolog 2 (Drosophila), Novel zinc finger protein, novelzin, Putative NF-kappa-B-activating protein 002N, SKD, Skeletrophin, Zinc finger ZZ type with ankyrin repeat domain protein 1, ZZ type with ankyrin repeat domain 1, ZZANK1
Gene Symbols MIB2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 925 and 975 of human mindbomb homolog 2 (Skeletrophin) using the numbering given in entry NP_543151.1 (GeneID 142678).
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 142678
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.