Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MINDY4B/FAM188B2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$451.65
Specifications
| Antigen | MINDY4B/FAM188B2 |
|---|---|
| Concentration | 0.5 mg/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MINDY4B/FAM188B2 Polyclonal antibody specifically detects MINDY4B/FAM188B2 in Human samples. It is validated for Western BlotSpecifications
| MINDY4B/FAM188B2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C3orf76, epididymis secretory sperm binding protein, FAM188B2, family with sequence similarity 188 member B2, inactive ubiquitin carboxyl-terminal hydrolase MINDY-4B, protein FAM188B2, Putative UPF0526 protein B | |
| The immunogen is a synthetic peptide directed towards the middle region of human MINDY4B/FAM188B2 (LOC646951): SQETLHGVLTRSDVGYLQWGKDASEDDRLSQVGSMLKTPKLPIWLCNING | |
| Affinity purified |
| 0.5 mg/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 1x PBS in 2% sucrose | |
| 646951 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title