Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Mini Electrolysis Device
SDPSCI_ED

Catalog No. s07375 Shop All United Scientific Supplies, Inc Products
Click to view available options
Class Size:
1 Lab Group

Includes

Activity guide and an instructional CD

The unique design of the Mini Electrolysis Device helps correct issues such as falling test tubes perched over unsteady electrodes and gases escaping due to poor alignment.

  • Ideal for teaching concepts such as Mixtures and Compounds, States of Matter, Oxidation Reduction Reactions, Law of Thermodynamics and Alternative Energy.
  • Each device consists of the electrolysis unit, gas collection tubes and a clear polycarbonate tub.
  • The device will attach to any 6 to 12V DC power supply (not included) using the alligator clips on the unit's power leads.
  • Shipping weight: 1 lb.
  • Dimensions: 7 in. x 4.5 in. x 3.5 in.

Specifications

Grade 5+
For Use With (Application) Helps correct issues such as falling test tubes perched over unsteady electrodes and gases escaping due to poor alignment, collect the correct proportions of hydrogen and oxygen from the electrolysis of water
Includes Activity guide and an instructional cd
Class Size 1 Lab Group
Product Type Mini Electrolysis Device
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.