Learn More
3M™ Mini Flip-Top Vial, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. p-7232135 Shop All AquaPhoenix Scientific Products
Click to view available options
Media Type:
9 mL letheen broth
9 mL maximum recovery diluent
Buffered Peptone Water Broth
Leak-resistant plastic cylinder with flip-open lid that seals securely for safe shipping and handling
  • Hinged lid can be opened with one hand, stays open for easy sample additions
  • Decontaminated through gamma irradiation and ready for use with no additional blending required
  • Flat bottom tube stands on its own, no holder required
  • Leak resistant vial helps prevent leakage during shipping
  • 2 year shelf life from date of manufacture provides long-term readiness
Specifications
Cap Type Flip-top
Media Type Buffered Peptone Water Broth
Product Type Microbiology Media
Quantity 100/Cs.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.