Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Minocycline hydrochloride
GREENER_CHOICESDP

Catalog No. 326850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-685-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-685-0 Supplier R&D Systems™ Supplier No. 3268/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antibiotic. Displays neuroprotective effects

Minocycline hydrochloride is a tetracycline antibiotic. Displays neuroprotective, antiapoptotic, anti-inflammatory and antimicrobial effects. Acts as a matrix metalloproteinase (MMP) inhibitor; attenuates disease severity in mouse models of multiple sclerosis. Orally active and brain penetrant. Allows formation of extended pluripotent stem (EPS) cells in combination with CHIR 99021 (Cat.No. 4423), (S)-(+)-Dimethindene maleate (Cat.No. 1425) and human leukemia inhibitory factor.

For more information about how Minocycline hydrochloride may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .

Chemical Identifiers

CAS 13614-98-7
Molecular Formula C23H28ClN3O7

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 493.94
Purity >98%
Molecular Weight (g/mol) 493.94
Chemical Name or Material Minocycline hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.