Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MINPP1 Antibody (OTI1B2), PE, Novus Biologicals™
SDP

Catalog No. NB287870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287870 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287870 Supplier Novus Biologicals Supplier No. NBP272693PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MINPP1 Monoclonal antibody specifically detects MINPP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MINPP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI1B2
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 3.1.3.62, HIPER1, Inositol (13,4,5)-tetrakisphosphate 3-phosphatase, Ins(13,4,5)P(4) 3-phosphatase, MINPP2, MIPPDKFZp564L2016, multiple inositol polyphosphate histidine phosphatase, 1, multiple inositol polyphosphate phosphatase 1, multiple inositol polyphosphate phosphatase 2, multiple inositol-polyphosphate phosphatase 1
Host Species Mouse
Immunogen Full length human recombinant protein of human MINPP1(NP_004888) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9562
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.