Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MIRA-1
GREENER_CHOICE

Catalog No. 336210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-6210 10 mg
33-625-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-6210 Supplier Tocris Bioscience™ Supplier No. 3362/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Restores mutant p53 activity; proapoptotic

Restores wild-type conformation, function and DNA binding activity to mutant p53. Induces p53 transcriptional transactivation of p21, MDM2 and PUMA, and promotes tumor cell death by apoptosis in a mutant p53-dependent manner in vitro (IC50 = 10μM).

Chemical Identifiers

CAS 72835-26-8
Molecular Formula C8H9NO4

Specifications

Quantity 10 mg
Formula Weight 183.16
Purity >98%
Molecular Weight (g/mol) 183.163
Chemical Name or Material MIRA-1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.