Learn More
United Scientific™ Mirrors, Flexible Acrylic, Double Sided
Shop All United Scientific Supplies, Inc Products
Click to view available options
Material:
Acrylic
Description
- Set of 12 double-sided acrylic mirrors in assorted sizes. Flexible design, ideal for demonstrating concave and convex properties.
- Contains four 7.5 x 6 in. mirrors, four 4 x 3 in. mirrors, and four 3 x 2 in. mirrors
- Mirror supports not included
- Material can be cut easily with scissors
Specifications
Specifications
| Material | Acrylic |
| Product Type | Mirror Assortment |
| Shape | Various |