Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT™-293 Transfection Reagent
Shop All Mirus Bio LLC Products

Click to view available options
Quantity:
1 x 0.4 mL
1 x 1 mL
5 x 1 mL
10 x 1 mL
Description
- Optimizes transfection performance in HEK 293 cells
- Low cellular toxicity maintains cell density and reduces experimental bias
- Allows 75 to 85% transfection efficiency in HEK 293 cell lines
- Delivers single or multiple plasmid for specialized applications such as virus production
- Supplied as a ready-to-use solution
- Concentration: 1.41 mg/mL
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Store for up to one year at 4°C (do not freeze). |
| Cell Type | 293-derived Cells |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Concentration | 1.41 mg/mL |
| For Use With (Application) | High efficiency, low-toxicity DNA transfection in HEK293 cells |
| Product Type | Transfection Reagent |
| Quantity | 1 x 0.4 mL |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction