Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-Jukart Transfection Reagent

Catalog No. p-2797906 Shop All Mirus Bio LLC Products
Click to view available options
Quantity:
5 x 1 mL
10 x 1 mL

Includes

TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution

TransIT™-Jukart Transfection Reagent optimized to achieve 10 – 25% transfection efficiency in Jurkat cells.

  • TransIT-Jurkat Transfection Reagent supplied as ready-to-use solution.
  • Cells can be transfected in serum-containing media, eliminating the need for media changes and improving transfection effeciency.
  • Concentration: 10 mg/mL

Specifications

Cell Type Primary Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 10 mg/mL
For Use With (Application) Gene expression and promoter analysis, optimized for Jurkat cells and other hard-to-transfect cells
Product Type Transfection Reagent
Quantity 5 x 1 mL
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.