Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT™-mRNA Transfection Kit
Shop All Mirus Bio LLC Products
Click to view available options
Quantity:
0.4 mL
1 mL
5 mL
10 mL
Description
Includes
TransIT-mRNA Transfection Reagent; mRNA Boost Reagent
- Low Cellular Toxicity: Maintain cell density and reduce experimental biases
- High Efficiency Delivery: Achieve RNA delivery in large population of cells to ensure experimental success
- Serum Compatible: Perform transfections in presence of serum which eliminates need for media change and maintains cellular health
- Deliver Various Sizes of RNA: Ideal for specialized applications, such as viral production and protein expression from mRNA
Specifications
Specifications
| Cell Type | Mammalian |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| For Use With (Application) | High efficiency delivery of both mRNA and viral RNA to various types of mammalian cells |
| Includes | 5 mL TransIT™-mRNA Transfection Reagent, 5 mL mRNA Boost Reagent |
| Product Type | Transfection Reagent |
| Quantity | 5 mL |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction