Learn More
Description
Specifications
Specifications
| Antigen | MIS/AMH |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3 |
| Conjugate | Biotin |
| Dilution | ELISA, Sandwich ELISA Detection |
| Formulation | PBS |
| Gene Alias | MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human MIS/AMH (NP_000470.2; Leu25-Arg560). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
