Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIS/AMH Antibody (RM0288-8D29), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP211801C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211801C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211801C Supplier Novus Biologicals Supplier No. NBP211801C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

MIS/AMH Monoclonal specifically detects MIS/AMH in Mouse samples. It is validated for Western Blot.

Specifications

Antigen MIS/AMH
Applications Western Blot
Classification Monoclonal
Clone RM0288-8D29
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance
Gene Symbols AMH
Host Species Rat
Immunogen Mouse recombinant MIS/AMH
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 268
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.