Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIS/AMH Goat anti-Mouse, Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Specifications
Antigen | MIS/AMH |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
MIS/AMH Polyclonal antibody AF1446SP Specifically detects MIS/AMH in Mouse, Rat samples. It is validated for Immunohistochemistry, Western BlottingSpecifications
MIS/AMH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse, Rat | |
AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
AMH | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
P49000 | |
268 | |
E. coli-derived recombinant rat MIS/AMH Lys447-Arg553 Accession # P49000 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse and rat MIS/AMH in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human MIS is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title