Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIS12 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292369 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292369 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292369 Supplier Novus Biologicals Supplier No. NB1002358PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MIS12 Polyclonal antibody specifically detects MIS12 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen MIS12
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias 2510025F08Rik, hMis12, homolog of yeast Mis12, KNTC2AP, MGC2488, MIS12 homolog, MIS12, MIND kinetochore complex component, homolog (S. pombe), MIS12, MIND kinetochore complex component, homolog (yeast), MTW1, protein MIS12 homolog
Host Species Rabbit
Immunogen Maps to a region between residue 1 and 50 of human MIS12 homolog using the numbering given in entry NP_076944.1 (GeneID 79003).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 79003
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.