Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mis12 Complex Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE2585
Description
Anti-Mis12 Complex Antibody is a highly specific rabbit polyclonal antibody that targets Mis12 complex and has been tested in Immunocytochemistry, Immunofluorescence, and Western Blotting.
Specifications
Mis12 Complex | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This polyclonal antibody detects Mis12 complex in human. It will also detect individual subunits (Mis12, PMF1, Dsn1, Nsl1) in Western Blotting. | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG |
Immunocytochemistry, Western Blot, Immunofluorescence | |
Please refer to lot specific datasheet. | |
MIS12,PMF1,DSN1,NSL1 | |
Four subunits from human Mis12 complex. | |
100 μg | |
Epigenetics and Nuclear Function | |
NP_076944,NP_009152,NP_079194,NP_056286 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction