Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mis12 Complex Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $437.74
Specifications
Antigen | Mis12 Complex |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE2585-25UG
|
MilliporeSigma
ABE258525UG |
25 μg |
Each for $192.35
|
|
|||||
ABE2585MI
|
MilliporeSigma
ABE2585 |
100 μg |
Each for $437.74
|
|
|||||
Description
Anti-Mis12 Complex Antibody is a highly specific rabbit polyclonal antibody that targets Mis12 complex and has been tested in Immunocytochemistry, Immunofluorescence, and Western Blotting.Specifications
Mis12 Complex | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Epigenetics and Nuclear Function | |
MIS12,PMF1,DSN1,NSL1 | |
IgG | |
Affinity Purified |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Four subunits from human Mis12 complex. | |
Primary | |
This polyclonal antibody detects Mis12 complex in human. It will also detect individual subunits (Mis12, PMF1, Dsn1, Nsl1) in Western Blotting. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title