Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIST1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341516 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341516 Supplier Novus Biologicals Supplier No. NBP282008PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MIST1 Polyclonal antibody specifically detects MIST1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen MIST1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias basic helix-loop-helix domain containing, class B, 8, basic helix-loop-helix family, member a15, bHLHa15, bHLHb8, class A basic helix-loop-helix protein 15, Class B basic helix-loop-helix protein 8, class II bHLH protein MIST1, MIST1MIST-1, Muscle, intestine and stomach expression 1
Host Species Rabbit
Immunogen MIST1 antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human MIST1. Amino Acid Squence: ATEAQPQGHLQRYSTQI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168620
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.