Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MITD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | MITD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MITD1 Polyclonal specifically detects MITD1 in Human samples. It is validated for Western Blot.Specifications
MITD1 | |
Polyclonal | |
Rabbit | |
Q8WV92 | |
129531 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
LOC129531, MIT domain-containing protein 1, MIT, microtubule interacting and transport, domain containing 1 | |
MITD1 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title