Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MITF Antibody (21D1418), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB10056561U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056561U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056561U Supplier Novus Biologicals Supplier No. NB10056561UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MITF Monoclonal specifically detects MITF in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MITF
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 21D1418
Conjugate DyLight 350
Dilution Western Blot
Gene Accession No. O75030
Gene Alias BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A
Gene Symbols MITF
Host Species Mouse
Immunogen A synthetic peptide mapping to the 400-450 a.a. region of accession number O75030.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4286
Test Specificity Mitf produces, by alternative splicing, 10 different transcripts, all with introns, putatively encoding 10 different protein isoforms. Therefore, observed molecular weights may vary depending on MITF isoforms expressed. Users are encouraged to utilize the peptide immunogen sequence to gain further information about the isoforms recognized by the antibody.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.