Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MITF Antibody (C5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11010872 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11010872 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11010872 Supplier Novus Biologicals Supplier No. NB11010872
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MITF Monoclonal specifically detects MITF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MITF
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2 ug/mg protein lysate, Immunohistochemistry-Paraffin 1 ug/ml
Gene Accession No. O75030
Gene Alias BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A
Gene Symbols MITF
Host Species Mouse
Immunogen N-terminal fragment of human microphthalmia protein.
Molecular Weight of Antigen 52-56 kDa
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4286
Test Specificity Microphthalmia (C5) this antibody recognizes the 52/56 kD band and not the longer 60-70 kD proteins. Mouse monoclonal clone C5 anti-Microphthalmia antibody recognizes serine phosphorylated and non-phosphorylated melanocytic isoforms of microphthalmia from human, mouse or rat.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.