Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MITF Antibody (C5/D5 Cocktail), Novus Biologicals™
SDP

Catalog No. NBP266627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266627 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266627 Supplier Novus Biologicals Supplier No. NBP266627
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MITF Monoclonal specifically detects MITF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays.

Specifications

Antigen MITF
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Shift
Classification Monoclonal
Clone C5/D5 Cocktail
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry, Immunoprecipitation 2 ug/mg protein lysate, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10 - 1:500, Gel Super Shift Assays
Formulation PBS with 0.08% Sodium Azide
Gene Accession No. O75030
Gene Alias BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A
Gene Symbols MITF
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from RBF/DnJ mice immunized with an N-terminal fragment of human microphthalmia protein and mouse myeloma NS1 cells.
Purification Method Protein A or G purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4286
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.