Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MITF Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
$110.70 - $492.00
Specifications
| Antigen | MITF |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ChIP sequencing (ChIP-seq) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3862
![]()
|
Active Motif
91202 |
10 μg |
Each for $110.70
|
|
|||||
50-199-3861
![]()
|
Active Motif
91201 |
100 μg |
Each for $492.00
|
|
|||||
Description
Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52 kDa and 56 kDa, while the longer Mi form runs as a cluster of bands at 60-70 kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.Specifications
| MITF | |
| ChIP sequencing (ChIP-seq) | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75030 | |
| 4286 | |
| This antibody was raised against an N-terminal fragment of human MITF (Micropthalmia Transcription Factor). | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| MITF |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 140mM HEPES, 35mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
| BCC2; BHLHE32; black eyed white; BOS_20839; Bw; Class E basic helix-loop-helix protein 32; CMM8; Gsfbcc2; melanocyte inducing transcription factor; melanogenesis associated transcription factor; Mi; microphtalmia-associated transcription factor; microphthalmia transcription factor; Microphthalmia-associated transcription factor; microphthalmia-associated transcription factor A; microphthalmia-associated transcription factor isoform H protein; microphthalmia-associated transcription factor Trnc-1; microphthalmia-associated transcription factor Trnc-2; microphthalmia-associated transcription factor variant 1; microphthalmia-associated transcription factor variant 2; microphthalmia-associated transcription factor variant 3; microphthalmia-associated transcription factor variant MITF-M(+); MITF; MITF-H; MITF-M; transcription factor, isoform 7; Vit; Vitiligo; Wh; WS2; WS2A | |
| MITF | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title