Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Mithramycin A
GREENER_CHOICESDP

Catalog No. 14891R Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-891-R 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-891-R Supplier Tocris Bioscience™ Supplier No. 1489/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of DNA and RNA polymerase

Mithramycin A is an anticancer antibiotic that selectively binds to G-C-rich DNA in the presence of Mg2+ or Zn2+, inhibiting RNA and DNA polymerase action. Inhibits c-myc expression and induces myeloid differentiation of HL-60 promyelocytic leukemia cells.

Chemical Identifiers

CAS 18378-89-7
Molecular Formula C52H76O24

Specifications

Quantity 1 mg
Solubility Soluble to 50mM in DMSO
Formula Weight 1085.16
Product Type Mithramycin A
Purity >95%
Molecular Weight (g/mol) 1085.156
Chemical Name or Material Mithramycin A
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.