Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitochondria Antibody (MTC02/2860R), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB379938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379938 Supplier Novus Biologicals Supplier No. NBP308548AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Mitochondria Monoclonal antibody specifically detects Mitochondria in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Mitochondria
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MTC02/2860R
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Semi-Purified mitochondrial preparation
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Mitochondrial Markers
Primary or Secondary Primary
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.