Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitochondrial ribosomal protein L11 Antibody (OTI1E5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB287869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287869 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287869 Supplier Novus Biologicals Supplier No. NBP272707APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mitochondrial ribosomal protein L11 Monoclonal antibody specifically detects Mitochondrial ribosomal protein L11 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Mitochondrial ribosomal protein L11
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E5
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias L11MT, MGC111024, mitochondrial ribosomal protein L11, MRP-L11,39S ribosomal protein L11, mitochondrial
Host Species Mouse
Immunogen Full length human recombinant protein of human MRPL11(NP_057134) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 65003
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.