Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mitochondrial ribosomal protein L30 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB143957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB143957 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB143957 Supplier Novus Biologicals Supplier No. NBP305921JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mitochondrial ribosomal protein L30 Polyclonal specifically detects mitochondrial ribosomal protein L30 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen mitochondrial ribosomal protein L30
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 39S ribosomal protein L30, mitochondrial, L28mt, L30mt, mitochondrial ribosomal protein L30, MRP-L28FLJ44438, MRPL28MGC3314,39S ribosomal protein L28, mitochondrial, MRP-L30, RPML28MGC24095
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human mitochondrial ribosomal protein L30 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51263
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.