Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitocondrial Translational Initiation Factor 3 Antibody (OTI4B6), HRP, Novus Biologicals™
SDP

Catalog No. NBP272708H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272708H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272708H Supplier Novus Biologicals Supplier No. NBP272708H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mitocondrial Translational Initiation Factor 3 Monoclonal specifically detects Mitocondrial Translational Initiation Factor 3 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Mitocondrial Translational Initiation Factor 3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4B6
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias FLJ33676, IF3(mt), IF-3mt, mitochondrial, mitochondrial translational initiation factor 3
Gene Symbols MTIF3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 56-278 of human Mitocondrial Translational Initiation Factor 3 (NP_690876) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 219402.0
Target Species Human, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.