Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitofilin Rabbit anti-Human, Clone: S03-6H5, Novus Biologicals™
SDP

Catalog No. NB188472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
25 ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188472 25 ul
NB159289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188472 Supplier Novus Biologicals Supplier No. NBP31506025UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Mitofilin Monoclonal antibody specifically detects Mitofilin in Human samples. It is validated for Western Blot

Specifications

Antigen Mitofilin
Applications Western Blot
Classification Monoclonal
Clone S03-6H5
Conjugate Unconjugated
Dilution Western Blot
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias DKFZp779P1653, inner membrane protein, mitochondrial, MGC111146, Mic 60, Mic60, MICOS Complex Subunit MIC60, mitochondrial (mitofilin), mitofilin, motor protein, p87/89, proliferation-inducing gene 4
Host Species Rabbit
Immunogen A synthetic peptide of human Mitofilin (Uniprot#: Q16891)
Purification Method Affinity purified
Quantity 25 ul
Regulatory Status RUO
Research Discipline Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10989
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.