Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitofilin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093180 100 μL
NB093181 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093180 Supplier Novus Biologicals Supplier No. NBP303850100UL

Rabbit Polyclonal Antibody

Mitofilin Polyclonal specifically detects Mitofilin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Mitofilin
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZp779P1653, inner membrane protein, mitochondrial, MGC111146, Mic 60, Mic60, MICOS Complex Subunit MIC60, mitochondrial (mitofilin), mitofilin, motor protein, p87/89, proliferation-inducing gene 4
Host Species Rabbit
Immunogen Recombinant protein of human Mitofilin.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10989
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.