Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitofusin 1 Antibody (11E9-1H12), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP171775UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171775UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171775UV Supplier Novus Biologicals Supplier No. NBP171775UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mitofusin 1 Monoclonal specifically detects Mitofusin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen Mitofusin 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 11E9-1H12
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
Gene Alias DKFZp762F247, EC 3.6.5, EC 3.6.5.-, FLJ20693, Fzo homolog, hfzo1, hfzo2, MGC41806, mitochondrial transmembrane GTPase Fzo-1, mitochondrial transmembrane GTPase FZO-2, mitofusin 1, mitofusin-1, putative transmembrane GTPase, Transmembrane GTPase MFN1
Gene Symbols MFN1
Host Species Mouse
Immunogen Mouse Mitofusin 1 [Swiss-Prot: Q811U4]
Molecular Weight of Antigen 80 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Mitochondrial Fusion Proteins, Mitochondrial Markers, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55669
Test Specificity Human and Mouse. Rat reactivity reported in scientific liteRature (PMID: 23658809).
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.