Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitofusin 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB338234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338234 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338234 Supplier Novus Biologicals Supplier No. NBP151841MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mitofusin 1 Polyclonal antibody specifically detects Mitofusin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Mitofusin 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp762F247, EC 3.6.5, EC 3.6.5.-, FLJ20693, Fzo homolog, hfzo1, hfzo2, MGC41806, mitochondrial transmembrane GTPase Fzo-1, mitochondrial transmembrane GTPase FZO-2, mitofusin 1, mitofusin-1, putative transmembrane GTPase, Transmembrane GTPase MFN1
Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of human Mitofusin-1 (within residues 5-100). [Swiss-Prot Q8IWA4]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Mitochondrial Fusion Proteins, Mitochondrial Markers, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55669
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.