Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MitoPY1
GSA_VAGREENER_CHOICESDP

Catalog No. 442810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-281-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-281-0 Supplier Tocris Bioscience™ Supplier No. 4428/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fluorescent mitochondrial hydrogen peroxide indicator

Fluorescent probe for imaging hydrogen peroxide (H2O2) in mitochondria of living cells. Displays enhanced emission at 528 nm in the presence of H2O2. Selective for H2O2 over superoxide, nitric oxide and hydroxyl radical. Detects local increases in H2O2 in an in vitro model of Parkinson's Disease.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1041634-69-8
Molecular Formula C52H53BIN2O5P

Specifications

Quantity 10 mg
Formula Weight 954.68
Purity >98%
Molecular Weight (g/mol) 954.693
Chemical Name or Material MitoPY1
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.