Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIXL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094229 20 μL
NB094228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094229 Supplier Novus Biologicals Supplier No. NBP30415920UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MIXL1 Polyclonal specifically detects MIXL1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MIXL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias hMix, homeobox protein MIXL1, Homeodomain protein MIX, MGC138179, MILD1, Mix.1 homeobox-like protein, Mix1 homeobox (Xenopus laevis)-like 1, Mix1 homeobox-like 1 (Xenopus laevis), MIX1 homeobox-like protein 1, Mix-like homeobox protein 1, MIXLMIX
Host Species Rabbit
Immunogen A synthetic peptide of human MIXL1.
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83881
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.