Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIXL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MIXL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MIXL1 Polyclonal specifically detects MIXL1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MIXL1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
83881 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
hMix, homeobox protein MIXL1, Homeodomain protein MIX, MGC138179, MILD1, Mix.1 homeobox-like protein, Mix1 homeobox (Xenopus laevis)-like 1, Mix1 homeobox-like 1 (Xenopus laevis), MIX1 homeobox-like protein 1, Mix-like homeobox protein 1, MIXLMIX | |
A synthetic peptide of human MIXL1. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title