Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MK 2206 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
MK 2206 dihydrochloride is a potent and selective allosteric Akt inhibitor (IC50 values are 5 nM, 12 nM, and 65 nM for Akt1, Akt2, and Akt3, respectively). MK 2206 requires the Pleckstrin homology domain for its activity, and exhibits no inhibitory activity in a panel of 250 tested protein kinases. MK 2206 induces growth inhibition of different cancer cell lines (IC50 in the range 3.4 and 28.6 μmol/L) and enhances antitumor efficacy of a range of standard chemotherapeutics. It synergistically inhibits cell proliferation of human lung and breast cancer cells in combination with Erlotinib (Cat. No. 7194) by suppressing both the Ras/Erk and PI 3-K pathways in vitro and in vivo. MK 2206 can be used to block the IL-22 - Akt signaling pathway for studying paneth cell differentiation.
Specifications
Specifications
| Chemical Name or Material | 8-[4-(1-Aminocyclobutyl)phenyl]-9-phenyl-1,2,4-triazolo[3,4-f][1,6]naphthyridin-3(2H)-one dihydrochloride |
| CAS | 1032350-13-2 |
| Quantity | 10 mg |
| Target | Akt (Protein Kinase B) Inhibitors |
| Molecular Formula | C25H21N5O.2HCl |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction