Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MK 386
GREENER_CHOICESDP

Catalog No. 58421 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-421 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 58-421 Supplier R&D Systems™ Supplier No. 5842/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective human type 1 5α-reductase inhibitor

Potent and selective human type 1 5α-reductase inhibitor (5αR1, IC50 = 0.9 nM). Exhibits 100-fold selectivity for 5αR1 over 5αR2 (IC50 = 154 nM). Supresses human serum and sebum dihydrotestosterone (DHT) levels in a dose-dependent manner without affecting semen DHT concentrations. Orally active.

Chemical Identifiers

CAS 158493-17-5
Molecular Formula C28H49NO

Specifications

Quantity 1 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Formula Weight 415.69
Physical Form Oil
Purity >98%
Molecular Weight (g/mol) 415.706
Chemical Name or Material (4aR,4bS,6aR,7R,9aS,9bS,10S,11aR)-7-[(1R)-1,5-Dimethylhexyl]hexadecahydro-1,4a,6a,10-tetramethyl-2H-
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.