Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MK 571
GREENER_CHOICESDP

Catalog No. 233810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-381-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-381-0 Supplier Tocris Bioscience™ Supplier No. 2338/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CysLT1 (LTD4) inverse agonist. Also MRP1 inhibitor

Potent CysLT1 (LTD4) leukotriene receptor inverse agonist (EC50 = 1.3 nM). Antagonizes LTD4-induced contractions of guinea pig trachea and ileum (pA2 values are 9.4 and 10.5 respectively). Also inhibitor of multidrug resistance protein-1 (MRP1) mediated transport; in vitro augments the effects of cytotoxic agents on malignant cells.

Chemical Identifiers

CAS 115104-28-4
Molecular Formula C26H27ClN2O3S2

Specifications

Quantity 10 mg
Formula Weight 515.09
Purity >98%
Molecular Weight (g/mol) 515.083
Chemical Name or Material MK 571
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.