Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKK3/MEK3 Rabbit anti-Human, Clone: JJ087-09, Novus Biologicals™
SDP

Catalog No. NBP267873 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267873 Supplier Novus Biologicals Supplier No. NBP267873

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

MKK3/MEK3 Monoclonal specifically detects MKK3/MEK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MKK3/MEK3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JJ087-09
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias MAP kinase kinase 3, MAPK/ERK kinase 3, MAPKK 3, MAPKK3PRKMK3dual specificity mitogen-activated protein kinase kinase 3, MEK3MEK 3, mitogen activated protein kinase kinase 3, mitogen-activated protein kinase kinase 3, MKK3EC 2.7.12.2
Gene Symbols MAP2K3
Host Species Rabbit
Immunogen Synthetic peptide within Human MKK3/MEK3 aa 298-347/ 347. (SwissProt: P46734 Human; SwissProt: P52564 Human; SwissProt: O09110 Mouse; SwissProt: P70236 Mouse; Entrez Gene: 114495 Rat; Entrez Gene: 303200 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Angiogenesis, Cancer, Hypoxia, Lipid and Metabolism, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5606
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.